Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039667.2 | NP_001034756.1 | 1 | 318 | ||
NM_139314.2 | NP_647475.1 | 1 | 318 | |||
NR_104213.1 | 1 | 318 | ||||
XM_005272484.3 | XP_005272541.1 | 1 | 316 | |||
XM_005272485.3 | XP_005272542.1 | 1 | 316 | |||
GenBank mRNA | AB056477.1 | - | 1 | 292 | ||
AF153606.1 | - | 1 | 276 | |||
AF169312.1 | - | 1 | 187 | |||
AF202636.1 | - | 1 | 295 | |||
AK222489.1 | - | 1 | 290 | |||
AK222524.1 | - | 1 | 288 | |||
AK303269.1 | - | 1 | 289 | |||
AY358275.1 | - | 1 | 286 | |||
BC023647.2 | - | 1 | 262 | 5088323 | ||
BG424907.1 | - | 1 | 318 |