Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015404.1 | 1 | 158 | |||
GenBank mRNA | AK096935.1 | - | 1 | 158 | ||
BC136540.1 | - | 1 | 83 | |||
JQ794500.1 | - | 1 | 261 |
Gene Symbol: MAPKAPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003668.3 | NP_003659.2 | 1 | 514 | ||
NM_139078.2 | NP_620777.1 | 1 | 514 | |||
XM_017020126.1 | XP_016875615.1 | 1 | 127 | |||
XM_017020127.1 | XP_016875616.1 | 1 | 437 | |||
XM_017020128.1 | XP_016875617.1 | 1 | 437 | |||
XM_017020129.1 | XP_016875618.1 | 1 | 274 | |||
XM_017020131.1 | XP_016875620.1 | 1 | 437 | |||
XM_017020132.1 | XP_016875621.1 | 1 | 372 | |||
XM_017020133.1 | XP_016875622.1 | 1 | 268 | |||
XM_017020135.1 | XP_016875624.1 | 1 | 267 | |||
XM_017020136.1 | XP_016875625.1 | 1 | 268 | |||
XM_017020137.1 | XP_016875626.1 | 1 | 267 | |||
XM_017020139.1 | XP_016875628.1 | 1 | 267 | |||
GenBank mRNA | AK122767.1 | - | 1 | 764 | ||
AW780199.1 | - | 1 | 268 | |||
BC041049.1 | - | 1 | 501 | 4909021 | ||
BC047284.1 | - | 1 | 182 | 4997201 | ||
BG502445.1 | - | 1 | 516 |