Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164276.1 | NP_001157748.1 | 5 | 1043 | ||
NM_016264.3 | NP_057348.3 | 4 | 899 | |||
XM_005259934.3 | XP_005259991.1 | 4 | 909 | |||
XM_011528062.2 | XP_011526364.1 | 5 | 1008 | |||
XM_011528064.1 | XP_011526366.1 | 4 | 1324 | |||
XM_011528066.1 | XP_011526368.1 | 3 | 1197 | |||
XM_017026870.1 | XP_016882359.1 | 4 | 1003 | |||
XM_017026871.1 | XP_016882360.1 | 5 | 1042 | |||
GenBank mRNA | AB021642.1 | - | 4 | 834 | ||
AK297528.1 | - | 5 | 957 | |||
BC136502.1 | - | 5 | 938 | 9020491 | ||
BE766488.1 | - | 1 | 393 |