Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018014.3 | NP_060484.2 | 3 | 656 | ||
NM_022893.3 | NP_075044.2 | 3 | 656 | |||
NM_138559.1 | NP_612569.1 | 3 | 656 | |||
XM_011532909.1 | XP_011531211.1 | 3 | 656 | |||
XM_011532910.1 | XP_011531212.1 | 3 | 656 | |||
XM_017004333.1 | XP_016859822.1 | 3 | 424 | |||
XM_017004336.1 | XP_016859825.1 | 3 | 219 | |||
XM_017004337.1 | XP_016859826.1 | 2 | 7125 | |||
XM_017004338.1 | XP_016859827.1 | 2 | 2869 | |||
GenBank mRNA | AB058712.1 | - | 3 | 556 | ||
AJ404611.1 | - | 3 | 656 | |||
AJ404612.1 | - | 3 | 656 | |||
AJ404613.1 | - | 3 | 656 | |||
AK055251.1 | - | 3 | 656 | |||
AK092316.1 | - | 2 | 95 | |||
AK300012.1 | - | 4 | 884 | |||
BC021098.1 | - | 3 | 537 | 5087967 |