Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142552.1 | NP_001136024.1 | 5 | 3787 | ||
NM_017639.3 | NP_060109.2 | 9 | 1920 | |||
XM_011532045.1 | XP_011530347.1 | 5 | 3840 | |||
XM_011532046.2 | XP_011530348.1 | 4 | 1496 | |||
XM_017008320.1 | XP_016863809.1 | 5 | 3840 | |||
GenBank mRNA | AK127704.1 | - | 5 | 3779 | ||
AY354497.1 | - | 9 | 1920 | |||
BC140919.1 | - | 5 | 3465 |