Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM208B SIRNA Assays ›
Gene Symbol:
FAM208B
Gene Name:
family with sequence similarity 208 member B
Gene Aliases:
C10orf18, bA318E3.2
Chromosome Location:
Chr.10: 5684759 - 5763740 on Build GRCh38
UniGene ID:
Hs.610717
Species:
Human
siRNA ID
HSS147809
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM208B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321783.1
NP_001308712.1
14
2968
NM_001321784.1
NP_001308713.1
14
3014
NM_001321785.1
NP_001308714.1
13
2725
NM_017782.4
NP_060252.4
13
2895
XM_005252474.3
XP_005252531.1
16
9706
XM_005252475.3
XP_005252532.1
17
3367
XM_005252476.4
XP_005252533.1
15
3150
XM_005252480.4
XP_005252537.1
13
2955
XM_005252481.4
XP_005252538.1
11
2440
XM_005252482.2
XP_005252539.1
15
2800
XM_011519515.2
XP_011517817.1
17
9844
XM_011519516.2
XP_011517818.1
17
9844
XM_011519517.2
XP_011517819.1
17
3764
XM_011519518.2
XP_011517820.1
16
3077
XM_011519519.2
XP_011517821.1
16
9601
XM_011519520.2
XP_011517822.1
14
9541
XM_011519521.2
XP_011517823.1
16
3288
XM_011519522.2
XP_011517824.1
16
3672
XM_011519525.2
XP_011517827.1
14
2708
XM_017016361.1
XP_016871850.1
16
9601
XM_017016362.1
XP_016871851.1
15
3571
XM_017016363.1
XP_016871852.1
16
3229
XM_017016364.1
XP_016871853.1
14
3433
XM_017016365.1
XP_016871854.1
15
3095
GenBank mRNA
AK128051.1
-
1
170
BX649177.1
-
5
1868
CR936780.1
-
13
2896
DB094755.1
-
1
542
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0