Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145265.1 | NP_001138737.1 | 3 | 1122 | ||
NM_001145266.1 | NP_001138738.1 | 3 | 1025 | |||
NM_018389.4 | NP_060859.4 | 2 | 1396 | |||
XM_011520202.2 | XP_011518504.1 | 2 | 283 | |||
GenBank mRNA | AF323970.1 | - | 2 | 684 | ||
AF326199.1 | - | 2 | 1293 | |||
AK002182.1 | - | 3 | 798 | |||
AK027394.1 | - | 2 | 1373 | |||
AK027747.1 | - | 2 | 1337 | |||
AK074929.1 | - | 2 | 1397 | |||
AK091245.1 | - | 2 | 2080 | |||
AK225428.1 | - | 3 | 820 | |||
AK315473.1 | - | 2 | 1098 | |||
BC001427.2 | - | 2 | 701 | 3139599 |