Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267571.1 | NP_001254500.1 | 7 | 1629 | ||
NM_001267572.1 | NP_001254501.1 | 1 | 408 | |||
NM_018421.3 | NP_060891.3 | 7 | 1629 | |||
NR_051978.1 | 6 | 1487 | ||||
XM_011518843.1 | XP_011517145.1 | 2 | 1511 | |||
XM_017014899.1 | XP_016870388.1 | 7 | 1059 | |||
GenBank mRNA | AB449882.1 | - | 7 | 1449 | ||
AF318370.1 | - | 7 | 1488 | |||
AK026105.1 | - | 6 | 977 | |||
AK124772.1 | - | 6 | 2178 | |||
AK131290.1 | - | 7 | 1507 | |||
AK298575.1 | - | 8 | 1279 | |||
AL353935.1 | - | 2 | 1690 | |||
AY026527.1 | - | 7 | 1541 | |||
BC028918.1 | - | 1 | 408 | 4931447 | ||
BC071978.1 | - | 3 | 516 | 5443205 |