Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017528.2 | NP_001017528.1 | 6 | 592 | ||
NM_001017529.2 | NP_001017529.1 | 5 | 475 | |||
NM_001017530.1 | NP_001017530.1 | 5 | 649 | |||
NM_001198721.1 | NP_001185650.1 | 7 | 1305 | |||
NM_015366.3 | NP_056181.2 | 6 | 766 | |||
NM_181333.3 | NP_851850.1 | 5 | 627 | |||
GenBank mRNA | AF177331.1 | - | 5 | 571 | ||
AK000192.1 | - | 1 | 685 | |||
AK055848.1 | - | 7 | 1028 | |||
AK091884.1 | - | 6 | 517 | |||
AK291564.1 | - | 5 | 617 | |||
BC016921.1 | - | 1 | 874 | 4413787 | ||
BI825567.1 | - | 5 | 631 |
Gene Symbol: PRR5-ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL355192.1 | - | 5 | 350 | ||
BI488517.1 | - | 6 | 425 |