Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290263.1 | NP_001277192.1 | 27 | 3109 | ||
NM_014689.2 | NP_055504.2 | 27 | 3252 | |||
XM_005246673.4 | XP_005246730.1 | 27 | 3111 | |||
XM_006712617.3 | XP_006712680.1 | 27 | 3276 | |||
XM_017004441.1 | XP_016859930.1 | 27 | 3279 | |||
XM_017004442.1 | XP_016859931.1 | 27 | 3282 | |||
XM_017004443.1 | XP_016859932.1 | 27 | 3328 | |||
XM_017004444.1 | XP_016859933.1 | 27 | 3280 | |||
XM_017004445.1 | XP_016859934.1 | 27 | 3109 | |||
XM_017004446.1 | XP_016859935.1 | 27 | 3280 | |||
XM_017004447.1 | XP_016859936.1 | 27 | 3278 | |||
GenBank mRNA | CR749492.1 | - | 2 | 97 | ||
EG328518.1 | - | 2 | 99 | |||
EU236710.2 | - | 27 | 3106 |