Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002860.3 | NP_001002860.2 | 3 | 1389 | ||
NM_018167.4 | NP_060637.1 | 3 | 1389 | |||
XM_011536939.2 | XP_011535241.1 | 3 | 1154 | |||
XM_017021438.1 | XP_016876927.1 | 1 | 154 | |||
GenBank mRNA | AB040958.2 | - | 3 | 1318 | ||
AK001510.1 | - | 3 | 1315 | |||
AK021953.1 | - | 1 | 608 | |||
AK303355.1 | - | 1 | 140 | |||
BC047071.1 | - | 3 | 1386 | |||
BC141850.1 | - | 3 | 1330 |