Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032946.2 | NP_116564.2 | 13, 14 | 1224 | ||
NR_028089.1 | 13, 14 | 1224 | ||||
GenBank mRNA | AJ277654.1 | - | 13, 14 | 1224 | ||
AJ277655.1 | - | 13, 14 | 1224 | |||
AJ277656.1 | - | 13, 14 | 1160 | |||
AJ277657.1 | - | 11, 12 | 1035 | |||
AJ277658.1 | - | 11, 12 | 1035 | |||
BC131708.1 | - | 12, 13 | 1045 | 40121743 |