Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028089.1 | 16, 17 | 1547 | |||
GenBank mRNA | AJ277654.1 | - | 16, 17 | 1547 | ||
AJ277655.1 | - | 16, 17 | 1522 | |||
AJ277656.1 | - | 16, 17 | 1458 | |||
AJ277657.1 | - | 14, 15 | 1333 | |||
AJ277658.1 | - | 14, 15 | 1452 |