Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018897.2 | NP_061720.2 | 42 | 7745 | ||
XM_011511487.2 | XP_011509789.1 | 43 | 8121 | |||
XM_011511488.2 | XP_011509790.1 | 41 | 8795 | |||
XM_011511489.2 | XP_011509791.1 | 41 | 7707 | |||
XM_011511490.2 | XP_011509792.1 | 41 | 8690 | |||
XM_011511491.2 | XP_011509793.1 | 42 | 8917 | |||
XM_011511492.2 | XP_011509794.1 | 42 | 8917 | |||
XM_011511493.2 | XP_011509795.1 | 42 | 8917 | |||
XM_011511494.2 | XP_011509796.1 | 42 | 8916 | |||
XM_011511495.2 | XP_011509797.1 | 42 | 8916 | |||
XM_011511497.2 | XP_011509799.1 | 9 | 2141 | |||
XM_017004504.1 | XP_016859993.1 | 41 | 8641 | |||
GenBank mRNA | AB023161.2 | - | 42 | 7763 | ||
AF327442.1 | - | 42 | 7740 |