Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278302.1 | NP_001265231.1 | 3 | 747 | ||
NM_001278303.1 | NP_001265232.1 | 3 | 775 | |||
NM_001278304.1 | NP_001265233.1 | 3 | 1377 | |||
NM_001278342.1 | NP_001265271.1 | 3 | 776 | |||
NM_019887.5 | NP_063940.1 | 5 | 1172 | |||
NM_138930.3 | NP_620308.1 | 4 | 879 | |||
GenBank mRNA | AF262240.1 | - | 4 | 373 | ||
AF298770.1 | - | 3 | 195 | |||
AK001399.1 | - | 3 | 1381 | |||
AK024768.1 | - | 4 | 381 | |||
AK057778.1 | - | 3 | 249 | |||
AK222664.1 | - | 4 | 370 | |||
AK315629.1 | - | 4 | 392 | |||
AY313210.1 | - | 3 | 222 | |||
BC004417.1 | - | 2 | 187 | |||
BC011909.2 | - | 4 | 377 | 4137792 | ||
BC046209.1 | - | 5 | 1094 | |||
BC095468.1 | - | 4 | 363 | 30557745 | ||
BF313712.1 | - | 3 | 249 | |||
BF724951.1 | - | 3 | 243 | |||
BM746689.1 | - | 3 | 218 | |||
CF128595.1 | - | 4 | 363 | |||
JX679472.1 | - | 2 | 63 | |||
KU178725.1 | - | 4 | 354 | |||
KU178726.1 | - | 3 | 222 |