Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A4RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020062.3 | NP_064446.2 | 3 | 395 | ||
GenBank mRNA | AB044777.1 | - | 2 | 286 | ||
AB052777.1 | - | 3 | 439 | |||
AK298304.1 | - | 1 | 318 | |||
BC001402.2 | - | 2 | 166 | |||
BC017446.1 | - | 1 | 177 | |||
BC052306.1 | - | 3 | 440 | 6576325 |