Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020243.4 | NP_064628.1 | 3, 4 | 387 | ||
GenBank mRNA | AB040119.1 | - | 3, 4 | 369 | ||
AB041906.1 | - | 3, 4 | 370 | |||
AF163256.1 | - | 3, 4 | 370 | |||
AK024731.1 | - | 3, 4 | 380 | |||
AK223021.1 | - | 3, 4 | 385 | |||
AK312076.1 | - | 3, 4 | 361 | |||
BC009363.2 | - | 3, 4 | 362 | 4302531 | ||
CR456475.1 | - | 3, 4 | 362 |