Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012329.1 | NP_001012329.1 | 3 | 307 | ||
NM_020248.2 | NP_064633.1 | 4 | 341 | |||
XM_017001853.1 | XP_016857342.1 | 4 | 566 | |||
XM_017001854.1 | XP_016857343.1 | 4 | 440 | |||
XM_017001855.1 | XP_016857344.1 | 3 | 283 | |||
GenBank mRNA | AB021262.1 | - | 4 | 307 | ||
BC014300.1 | - | 3 | 295 | |||
BC146892.1 | - | 1 | 70 | 9056820 | ||
BC146896.1 | - | 1 | 70 | 9056824 | ||
BI226022.1 | - | 294 | ||||
CB995652.1 | - | 334 |