Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012329.1 | NP_001012329.1 | 4 | 394 | ||
NM_020248.2 | NP_064633.1 | 5 | 428 | |||
XM_017001853.1 | XP_016857342.1 | 5 | 653 | |||
XM_017001854.1 | XP_016857343.1 | 5 | 527 | |||
XM_017001855.1 | XP_016857344.1 | 4 | 370 | |||
GenBank mRNA | AB021262.1 | - | 5 | 394 | ||
BC014300.1 | - | 4 | 382 | |||
BC146892.1 | - | 2 | 157 | 9056820 | ||
BC146896.1 | - | 2 | 157 | 9056824 | ||
BI226022.1 | - | 381 | ||||
CB995652.1 | - | 421 |