Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020451.2 | NP_065184.2 | 7 | 993 | ||
NM_206926.1 | NP_996809.1 | 6 | 891 | |||
GenBank mRNA | AF166125.1 | - | 5 | 582 | ||
AJ306399.1 | - | 7 | 993 | |||
BC021028.2 | - | 2 | 196 | |||
BQ217758.1 | - | 4 | 448 |