Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029884.1 | NP_001025055.1 | 3 | 551 | ||
XM_005267064.3 | XP_005267121.1 | 2 | 2630 | |||
XM_005267065.3 | XP_005267122.1 | 4 | 577 | |||
XM_005267066.2 | XP_005267123.1 | 2 | 1434 | |||
XM_006715521.1 | XP_006715584.1 | 4 | 804 | |||
XM_006715522.3 | XP_006715585.1 | 2 | 2671 | |||
XM_011535981.2 | XP_011534283.1 | 2 | 3427 | |||
XM_011535982.2 | XP_011534284.1 | 2 | 3427 | |||
XM_011535983.2 | XP_011534285.1 | 3 | 1852 | |||
XM_017011102.1 | XP_016866591.1 | 2 | 1436 | |||
GenBank mRNA | AK056300.1 | - | 2 | 450 | ||
AK293288.1 | - | 2 | 482 | |||
AK294798.1 | - | 2 | 605 | |||
AK307631.1 | - | 2 | 504 | |||
BC140864.1 | - | 1 | 288 | 9021732 | ||
BC151134.1 | - | 1 | 288 | 9057034 |