Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAHCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291324.1 | NP_001278253.1 | 25 | 6274 | ||
XM_011525063.2 | XP_011523365.1 | 26 | 6642 | |||
XM_017024898.1 | XP_016880387.1 | 25 | 6325 | |||
XM_017024899.1 | XP_016880388.1 | 24 | 6231 | |||
GenBank mRNA | AB040880.1 | - | 21 | 4025 |