Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142641.1 | NP_001136113.1 | 16 | 2314 | ||
XM_005266171.3 | XP_005266228.1 | 19 | 2545 | |||
XM_005266173.3 | XP_005266230.1 | 19 | 2540 | |||
XM_005266175.3 | XP_005266232.1 | 18 | 2479 | |||
XM_005266176.3 | XP_005266233.1 | 18 | 2421 | |||
XM_005266177.3 | XP_005266234.1 | 19 | 2414 | |||
XM_011534803.2 | XP_011533105.1 | 19 | 2604 | |||
XM_011534804.2 | XP_011533106.1 | 19 | 2601 | |||
XM_011534805.2 | XP_011533107.1 | 18 | 2535 | |||
XM_011534806.2 | XP_011533108.1 | 18 | 2538 | |||
XM_011534807.2 | XP_011533109.1 | 18 | 2480 | |||
XM_011534808.2 | XP_011533110.1 | 19 | 2473 | |||
XM_011534809.2 | XP_011533111.1 | 18 | 2416 | |||
XM_011534810.2 | XP_011533112.1 | 18 | 2394 | |||
XM_011534812.2 | XP_011533114.1 | 20 | 2426 | |||
GenBank mRNA | AB046765.2 | - | 16 | 2085 | ||
AF490258.1 | - | 9 | 1180 |