Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020911.1 | NP_065962.1 | 20 | 4030 | ||
XM_005250686.4 | XP_005250743.1 | 20 | 4428 | |||
XM_006716171.3 | XP_006716234.1 | 21 | 4521 | |||
XM_011516676.2 | XP_011514978.1 | 20 | 3936 | |||
XM_017012779.1 | XP_016868268.1 | 19 | 3735 | |||
GenBank mRNA | AB046770.3 | - | 18 | 2507 |