Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005260476.2 | XP_005260533.1 | 4 | 647 | ||
XM_006723841.2 | XP_006723904.1 | 2 | 293 | |||
XM_011528952.2 | XP_011527254.1 | 4 | 708 | |||
XM_011528953.2 | XP_011527255.1 | 3 | 588 | |||
GenBank mRNA | AJ297792.1 | - | 4 | 655 | ||
AK292443.1 | - | 4 | 655 | |||
AY422170.1 | - | 4 | 664 | |||
BC035639.1 | - | 4 | 553 | 5547220 |