Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021221.2 | NP_067044.2 | 1 | 13 | ||
GenBank mRNA | AJ245417.1 | - | 1 | 585 | ||
AJ315545.1 | - | 1 | 13 | |||
AK289364.1 | - | 1 | 797 | |||
HE864448.1 | - | 1 | 33 | |||
HE864449.1 | - | 1 | 33 | |||
HE864450.1 | - | 1 | 33 | |||
HE864451.1 | - | 1 | 33 |