Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052813.4 | NP_434700.2 | 2, 3 | 352 | ||
NM_052814.3 | NP_434701.1 | 2, 3 | 352 | |||
GenBank mRNA | AF311287.1 | - | 2, 3 | 313 | ||
AK024001.1 | - | 2, 3 | 332 | |||
AK292081.1 | - | 2, 3 | 304 | |||
AK295647.1 | - | 2, 3 | 191 | |||
AK309043.1 | - | 2, 3 | 317 | |||
BC008877.2 | - | 2, 3 | 327 | 4139529 | ||
BI524042.1 | - | 2, 3 | 337 | |||
BM148244.1 | - | 2, 3 | 352 |