Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031714.3 | NP_001026884.3 | 17 | 2663 | ||
NM_022489.3 | NP_071934.3 | 17 | 2663 | |||
XM_005268004.4 | XP_005268061.1 | 17 | 2750 | |||
XM_005268005.4 | XP_005268062.1 | 17 | 2721 | |||
XM_017021595.1 | XP_016877084.1 | 17 | 2721 | |||
GenBank mRNA | AK025709.1 | - | 8 | 626 | ||
AK225325.1 | - | 8 | 626 | |||
BC008756.2 | - | 7 | 571 | |||
BU538960.1 | - | 3 | 139 | |||
DA500952.1 | - | 5 | 324 | |||
DQ395338.1 | - | 10 | 924 | |||
DQ395339.1 | - | 10 | 924 | |||
DQ395340.1 | - | 10 | 924 |