Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIGYF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022574.4 | NP_072096.2 | 21 | 3635 | ||
XM_005250532.1 | XP_005250589.1 | 23 | 3416 | |||
XM_011516472.1 | XP_011514774.1 | 23 | 3416 | |||
XM_011516477.1 | XP_011514779.1 | 23 | 3413 | |||
XM_017012526.1 | XP_016868015.1 | 23 | 3416 | |||
XM_017012527.1 | XP_016868016.1 | 25 | 3314 | |||
XM_017012528.1 | XP_016868017.1 | 24 | 2926 | |||
XM_017012529.1 | XP_016868018.1 | 24 | 2933 | |||
XM_017012530.1 | XP_016868019.1 | 24 | 3187 | |||
XM_017012531.1 | XP_016868020.1 | 19 | 2439 | |||
GenBank mRNA | AF289554.1 | - | 3 | 440 | ||
AY176044.1 | - | 21 | 3836 | |||
BC018563.1 | - | 5 | 609 | |||
BC129991.1 | - | 15 | 1783 |