Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSWIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023072.2 | NP_075560.2 | 13 | 2735 | ||
XM_005260023.3 | XP_005260080.1 | 14 | 2897 | |||
XM_017027153.1 | XP_016882642.1 | 14 | 3168 | |||
XM_017027154.1 | XP_016882643.1 | 14 | 3100 | |||
XM_017027155.1 | XP_016882644.1 | 13 | 3001 | |||
XM_017027156.1 | XP_016882645.1 | 13 | 2672 | |||
GenBank mRNA | AK022283.1 | - | 2 | 518 | ||
AK024452.1 | - | 13 | 2735 | |||
AK299057.1 | - | 12 | 2273 |