Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161527.1 | NP_001154999.1 | 4 | 497 | ||
NM_024309.3 | NP_077285.3 | 4 | 783 | |||
GenBank mRNA | AF372839.1 | - | 4 | 783 | ||
AJ304866.1 | - | 4 | 730 | |||
AK026176.1 | - | 4 | 496 | |||
AK096296.1 | - | 4 | 774 | |||
BC002740.2 | - | 4 | 741 | 3632736 | ||
DA421921.1 | - | 4 | 498 |