Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304833.1 | NP_001291762.1 | 3 | 337 | ||
NM_001304834.1 | NP_001291763.1 | 3 | 575 | |||
NM_001304835.1 | NP_001291764.1 | 3 | 575 | |||
NM_001304836.1 | NP_001291765.1 | 3 | 575 | |||
NM_001304837.1 | NP_001291766.1 | 2 | 518 | |||
NM_001304838.1 | NP_001291767.1 | 3 | 575 | |||
NM_024623.2 | NP_078899.1 | 4 | 724 | |||
GenBank mRNA | AK023553.1 | - | 4 | 620 | ||
AK094820.1 | - | 3 | 320 | |||
AK095481.1 | - | 2 | 426 | |||
AK303095.1 | - | 2 | 241 | |||
BC009181.1 | - | 3 | 583 | |||
BC015605.2 | - | 3 | 526 | |||
BC071172.1 | - | 2 | 1322 | |||
BC098119.1 | - | 3 | 307 | 40024912 | ||
BC098293.1 | - | 3 | 307 | 40024914 | ||
BC101940.1 | - | 3 | 307 | 40024915 | ||
BC105637.1 | - | 3 | 307 | 40024913 | ||
BX640675.1 | - | 3 | 309 | |||
BX640734.1 | - | 3 | 342 | |||
CR457316.1 | - | 1 | 108 | |||
DB009535.1 | - | 3 | 337 | |||
DC404984.1 | - | 2 | 241 |