Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323679.1 | NP_001310608.1 | 4 | 666 | ||
NM_001323680.1 | NP_001310609.1 | 3 | 615 | |||
NM_001323681.1 | NP_001310610.1 | 3 | 606 | |||
NM_001323682.1 | NP_001310611.1 | 4 | 672 | |||
NM_001323683.1 | NP_001310612.1 | 4 | 666 | |||
NM_001323684.1 | NP_001310613.1 | 4 | 694 | |||
NM_001323685.1 | NP_001310614.1 | 3 | 335 | |||
NM_024958.2 | NP_079234.1 | 3 | 593 | |||
XM_006723630.3 | XP_006723693.1 | 3 | 1016 | |||
XM_011529360.1 | XP_011527662.1 | 3 | 299 | |||
XM_011529362.2 | XP_011527664.1 | 4 | 306 | |||
XM_011529363.2 | XP_011527665.1 | 3 | 284 | |||
XM_017028073.1 | XP_016883562.1 | 3 | 238 | |||
XM_017028074.1 | XP_016883563.1 | 3 | 292 | |||
XM_017028075.1 | XP_016883564.1 | 3 | 464 | |||
XM_017028076.1 | XP_016883565.1 | 3 | 647 | |||
GenBank mRNA | AK026982.1 | - | 3 | 206 | ||
AK290527.1 | - | 3 | 570 | |||
AL136915.1 | - | 2 | 188 | |||
BC001963.1 | - | 2 | 173 | 3454427 | ||
BP202782.1 | - | 4 | 287 | |||
BP251173.1 | - | 4 | 287 | |||
CD102518.1 | - | 3 | 224 | |||
CR533441.1 | - | 1 | 55 | |||
DB172225.1 | - | 3 | 335 |