Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281739.2 | NP_001268668.1 | 20 | 3649 | ||
NM_001281740.2 | NP_001268669.1 | 24 | 4225 | |||
NM_025135.4 | NP_079411.2 | 21 | 3700 | |||
XM_005258349.1 | XP_005258406.1 | 22 | 4018 | |||
XM_005258354.1 | XP_005258411.1 | 20 | 3568 | |||
XM_005258355.1 | XP_005258412.1 | 20 | 3568 | |||
XM_011526189.1 | XP_011524491.1 | 25 | 4267 | |||
XM_011526190.1 | XP_011524492.1 | 25 | 4267 | |||
XM_011526192.1 | XP_011524494.1 | 24 | 4192 | |||
XM_011526193.2 | XP_011524495.1 | 25 | 4267 | |||
XM_011526195.1 | XP_011524497.1 | 24 | 3970 | |||
XM_011526196.1 | XP_011524498.1 | 22 | 3742 | |||
XM_011526197.1 | XP_011524499.1 | 24 | 4192 | |||
XM_017026006.1 | XP_016881495.1 | 25 | 4267 | |||
XM_017026007.1 | XP_016881496.1 | 23 | 3905 | |||
XM_017026008.1 | XP_016881497.1 | 25 | 4267 | |||
XM_017026009.1 | XP_016881498.1 | 21 | 3682 | |||
XM_017026010.1 | XP_016881499.1 | 23 | 3911 | |||
GenBank mRNA | AB051482.1 | - | 14 | 2841 | ||
AB084087.1 | - | 20 | 3509 | |||
AK025950.1 | - | 6 | 795 | |||
AK091899.1 | - | 13 | 1751 | |||
AL834480.1 | - | 4 | 487 | |||
BC031518.1 | - | 17 | 3152 | |||
BC050670.1 | - | 5 | 571 | |||
BC081563.1 | - | 6 | 827 | |||
HM191478.1 | - | 24 | 4085 |