Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030943.3 | NP_112205.2 | 7, 8 | 789 | ||
XM_011537202.2 | XP_011535504.1 | 7, 8 | 1034 | |||
XM_011537203.2 | XP_011535505.1 | 7, 8 | 1290 | |||
GenBank mRNA | AF328788.1 | - | 7, 8 | 778 | ||
AK055802.1 | - | 6, 7 | 1502 |