Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030949.2 | NP_112211.1 | 4 | 615 | ||
XM_005267152.4 | XP_005267209.1 | 4 | 1873 | |||
GenBank mRNA | AF308297.1 | - | 4 | 488 | ||
AF407165.2 | - | 4 | 1117 | |||
AK096272.1 | - | 4 | 251 | |||
BC015773.1 | - | 4 | 591 | 4877511 | ||
DR003369.1 | - | 4 | 625 |