Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BGRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031286.3 | NP_112576.1 | 3 | 669 | ||
GenBank mRNA | AF247790.1 | - | 3 | 318 | ||
AF304163.1 | - | 3 | 278 | |||
AF466367.1 | - | 3 | 1215 | |||
AJ297915.1 | - | 3 | 306 | |||
AK024884.1 | - | 3 | 292 | |||
BC030135.2 | - | 3 | 438 | 4040656 | ||
FJ224343.1 | - | 3 | 1215 |