Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136044.1 | NP_001129516.1 | 3 | 547 | ||
NM_031434.3 | NP_113622.1 | 3 | 1010 | |||
GenBank mRNA | AK314035.1 | - | 3 | 775 | ||
AY037155.1 | - | 3 | 528 | |||
AY358481.1 | - | 3 | 506 | |||
AY603380.1 | - | 2 | 532 | |||
BC000936.1 | - | 3 | 547 | 3449979 | ||
BC033182.1 | - | 3 | 685 | 4586932 |