Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032023.3 | NP_114412.2 | 6, 7 | 675 | ||
XM_005271833.4 | XP_005271890.1 | 6, 7 | 725 | |||
XM_005271835.4 | XP_005271892.1 | 6, 7 | 675 | |||
XM_006718013.3 | XP_006718076.1 | 6, 7 | 770 | |||
XM_006718014.3 | XP_006718077.1 | 5, 6 | 2189 | |||
XM_011540255.2 | XP_011538557.1 | 5, 6 | 550 | |||
XM_017016754.1 | XP_016872243.1 | 5, 6 | 2315 | |||
XM_017016755.1 | XP_016872244.1 | 5, 6 | 2270 | |||
XM_017016756.1 | XP_016872245.1 | 6, 7 | 796 | |||
XM_017016757.1 | XP_016872246.1 | 5, 6 | 2341 | |||
XM_017016758.1 | XP_016872247.1 | 4, 5 | 2046 | |||
GenBank mRNA | AB209446.1 | - | 5, 6 | 2547 | ||
AF260335.1 | - | 6, 7 | 639 | |||
AK055763.1 | - | 4, 5 | 1683 | |||
AK092770.1 | - | 3, 4 | 1885 | |||
AK097272.1 | - | 1, 2 | 1953 | |||
AY216713.1 | - | 6, 7 | 675 | |||
AY216714.1 | - | 6, 7 | 635 | |||
AY216715.1 | - | 6, 7 | 702 | |||
AY216716.1 | - | 5, 6 | 583 | |||
BC032593.1 | - | 6, 7 | 657 | 5527647 | ||
BI601296.1 | - | 6, 7 | 678 |