Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206998.1 | NP_001193927.1 | 8 | 1355 | ||
NM_032173.3 | NP_115549.2 | 8 | 1148 | |||
XM_011530435.2 | XP_011528737.1 | 9 | 1344 | |||
XM_011530436.2 | XP_011528738.1 | 10 | 1399 | |||
XM_011530437.1 | XP_011528739.1 | 9 | 2069 | |||
XM_011530438.2 | XP_011528740.1 | 9 | 1585 | |||
XM_017028990.1 | XP_016884479.1 | 9 | 1840 | |||
GenBank mRNA | AB051436.1 | - | 8 | 1220 | ||
AK025710.1 | - | 3 | 542 | |||
AK096397.1 | - | 8 | 1148 | |||
BC009252.2 | - | 3 | 479 | |||
BC021570.1 | - | 3 | 507 | |||
BC069019.1 | - | 3 | 478 | |||
BC094857.1 | - | 3 | 478 | |||
CR456397.1 | - | 8 | 1128 |