Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC37A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287498.1 | NP_001274427.1 | 5 | 532 | ||
NM_032295.3 | NP_115671.1 | 5 | 532 | |||
NM_207113.2 | NP_996996.1 | 5 | 532 | |||
XM_011516626.2 | XP_011514928.1 | 7 | 1029 | |||
XM_011516627.2 | XP_011514929.1 | 7 | 1028 | |||
XM_017012712.1 | XP_016868201.1 | 7 | 1005 | |||
XM_017012713.1 | XP_016868202.1 | 7 | 1024 | |||
XM_017012714.1 | XP_016868203.1 | 7 | 1045 | |||
XM_017012715.1 | XP_016868204.1 | 7 | 1048 | |||
XM_017012716.1 | XP_016868205.1 | 7 | 1052 | |||
XM_017012717.1 | XP_016868206.1 | 7 | 1024 | |||
XM_017012718.1 | XP_016868207.1 | 7 | 1023 | |||
GenBank mRNA | AK074823.1 | - | 5 | 493 | ||
AK299276.1 | - | 5 | 476 | |||
AK300265.1 | - | 5 | 490 | |||
AL136583.1 | - | 5 | 439 | |||
BC028380.1 | - | 5 | 482 | 4811814 | ||
BC046567.1 | - | 5 | 484 | 5246993 | ||
CD101739.1 | - | 5 | 540 | |||
DB481623.1 | - | 4 | 423 |