Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032306.3 | NP_115682.1 | 1 | 574 | ||
XM_017027355.1 | XP_016882844.1 | 1 | 210 | |||
GenBank mRNA | AK223336.1 | - | 1 | 197 | ||
AK311956.1 | - | 1 | 213 | |||
AY358858.1 | - | 1 | 574 | |||
AY427650.2 | - | 1 | 574 | |||
BC004393.1 | - | 1 | 229 | 3635540 | ||
BX334356.2 | - | 1 | 491 |