Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134395.1 | NP_001127867.1 | 4 | 438 | ||
NM_001134396.1 | NP_001127868.1 | 4 | 419 | |||
NM_001318252.1 | NP_001305181.1 | 4 | 1322 | |||
NM_032350.5 | NP_115726.1 | 4 | 448 | |||
NR_134537.1 | 3 | 424 | ||||
XM_006715793.2 | XP_006715856.1 | 3 | 1062 | |||
XM_011515580.2 | XP_011513882.1 | 3 | 1062 | |||
XM_011515581.2 | XP_011513883.1 | 4 | 482 | |||
XM_011515582.2 | XP_011513884.1 | 4 | 433 | |||
XM_011515583.2 | XP_011513885.1 | 4 | 457 | |||
XM_011515584.2 | XP_011513886.1 | 4 | 1138 | |||
XM_017012719.1 | XP_016868208.1 | 3 | 1062 | |||
XM_017012720.1 | XP_016868209.1 | 4 | 442 | |||
XM_017012721.1 | XP_016868210.1 | 5 | 1361 | |||
XM_017012722.1 | XP_016868211.1 | 4 | 481 | |||
GenBank mRNA | AF495721.1 | - | 4 | 1278 | ||
BC006224.2 | - | 4 | 448 | 3941780 | ||
BF129057.1 | - | 424 | ||||
BM762287.1 | - | 3 | 368 | |||
BM763235.1 | - | 4 | 422 |