Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040160.2 | NP_001035250.1 | 1 | 115 | ||
NM_001040161.2 | NP_001035251.1 | 1 | 115 | |||
NM_001040162.2 | NP_001035252.1 | 1 | 115 | |||
NM_001040165.2 | NP_001035255.1 | 1 | 115 | |||
NM_001288710.1 | NP_001275639.1 | 1 | 115 | |||
NM_032366.4 | NP_115742.3 | 1 | 115 | |||
NR_109976.1 | 1 | 115 | ||||
NR_109977.1 | 1 | 115 | ||||
NR_109978.1 | 1 | 115 | ||||
NR_109979.1 | 1 | 115 | ||||
XM_011522713.2 | XP_011521015.1 | 1 | 113 | |||
XM_011522714.2 | XP_011521016.1 | 1 | 113 | |||
GenBank mRNA | AK057077.1 | - | 1 | 65 | ||
AK293046.1 | - | 1 | 51 | |||
AK294482.1 | - | 1 | 51 | |||
AK295628.1 | - | 1 | 51 | |||
AW027232.1 | - | 1 | 27 | |||
BC007207.1 | - | 1 | 97 | 4054799 | ||
BG612714.1 | - | 1 | 80 | |||
BM827705.1 | - | 1 | 50 | |||
BP276058.1 | - | 1 | 82 | |||
BQ921757.1 | - | 20 | ||||
BU539308.1 | - | 1 | 63 | |||
CK904040.1 | - | 1 | 19 | |||
DB499720.1 | - | 1 | 115 |