Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242832.1 | NP_001229761.1 | 9, 10 | 1119 | ||
NM_032389.4 | NP_115765.2 | 10, 11 | 1203 | |||
XM_005253166.1 | XP_005253223.1 | 11, 12 | 1698 | |||
XM_005253167.1 | XP_005253224.1 | 11, 12 | 1665 | |||
XM_005253168.2 | XP_005253225.1 | 11, 12 | 1623 | |||
XM_006718346.1 | XP_006718409.1 | 10, 11 | 1656 | |||
XM_011520405.1 | XP_011518707.1 | 10, 11 | 1539 | |||
XM_017018413.1 | XP_016873902.1 | 10, 11 | 1623 | |||
XM_017018414.1 | XP_016873903.1 | 10, 11 | 1019 | |||
XM_017018415.1 | XP_016873904.1 | 10, 11 | 971 | |||
GenBank mRNA | AB073358.1 | - | 9, 10 | 813 | ||
AK027482.1 | - | 10, 11 | 939 | |||
AK098324.1 | - | 5, 6 | 549 | |||
AK301785.1 | - | 9, 10 | 823 | |||
AK304541.1 | - | 10, 11 | 778 | |||
AK315992.1 | - | 4, 5 | 470 | |||
AL834337.1 | - | 10, 11 | 922 | |||
BC030148.2 | - | 10, 11 | 946 | 4558453 | ||
BC036823.1 | - | 10, 11 | 948 |