Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNFN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032488.3 | NP_115877.2 | 2, 3 | 167 | ||
XM_005259332.2 | XP_005259389.1 | 3, 4 | 272 | |||
XM_011527396.1 | XP_011525698.1 | 3, 4 | 267 | |||
XM_011527397.1 | XP_011525699.1 | 3, 4 | 213 | |||
GenBank mRNA | AB049591.2 | - | 2, 3 | 162 | ||
AK312080.1 | - | 2, 3 | 167 | |||
BC101197.1 | - | 3, 4 | 223 | 40021735 | ||
BC101198.2 | - | 2, 3 | 143 | 40021738 |