Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032517.4 | NP_115906.3 | 5 | 607 | ||
GenBank mRNA | AI393331.1 | - | 1 | 203 | ||
AY358694.1 | - | 5 | 552 | |||
BC021730.2 | - | 5 | 573 | 4825273 | ||
BG209290.1 | - | 2 | 118 |