Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026838.1 | 2 | 221 | |||
NR_026839.1 | 2 | 221 | ||||
NR_026840.1 | 2 | 221 | ||||
NR_026841.1 | 2 | 221 | ||||
NR_026842.1 | 2 | 221 | ||||
GenBank mRNA | AA707621.1 | - | 3 | 336 | ||
AA770014.1 | - | 3 | 342 | |||
AF321193.1 | - | 2 | 196 | |||
AF426256.1 | - | 2 | 144 | |||
AJ001904.1 | - | 2 | 181 | |||
AJ297914.2 | - | 2 | 199 | |||
AJ301615.2 | - | 2 | 199 | |||
AJ305085.2 | - | 2 | 199 | |||
AJ306839.2 | - | 2 | 199 | |||
AJ306840.2 | - | 2 | 199 | |||
AJ783421.1 | - | 2 | 199 | |||
BC015981.1 | - | 2 | 203 | |||
BC029353.1 | - | 2 | 221 | |||
BC070229.1 | - | 2 | 180 | |||
BC117304.1 | - | 2 | 153 | |||
BC117330.1 | - | 2 | 153 | |||
BC143995.1 | - | 2 | 153 |