Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005366.1 | NP_001005366.1 | 11 | 1844 | ||
NM_032590.4 | NP_115979.3 | 11 | 1634 | |||
XM_005253955.3 | XP_005254012.1 | 11 | 1844 | |||
XM_005253956.3 | XP_005254013.1 | 11 | 1441 | |||
XM_011538867.2 | XP_011537169.1 | 11 | 2887 | |||
XM_011538868.2 | XP_011537170.1 | 11 | 2883 | |||
XM_011538869.2 | XP_011537171.1 | 11 | 2885 | |||
GenBank mRNA | AB031230.1 | - | 1 | 93 | ||
AJ459424.1 | - | 11 | 1606 | |||
AK027440.1 | - | 1 | 23 | |||
AK127328.1 | - | 11 | 1844 | |||
AL133572.1 | - | 7 | 1147 | |||
BC115379.1 | - | 10 | 1494 | |||
BC115380.1 | - | 11 | 1573 | 40029132 | ||
BX648928.1 | - | 9 | 1366 |