Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CROCCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054737.1 | - | 1 | 690 | ||
AK090414.1 | - | 11 | 4326 |
Gene Symbol: ADAM33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092627.1 | - | 8 | 2044 |
Gene Symbol: ITPKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025304.1 | - | 1 | 2134 |